• USA Home
  • 4.90294 - 1-Ethyl-3-methylimidazolium diethylphosphate

EMAIL THIS PAGE TO A FRIEND
4.90294 EMD Millipore

1-Ethyl-3-methylimidazolium diethylphosphate

1-Ethyl-3-methylimidazolium diethylphosphate for synthesis. CAS 848641-69-0, pH 5 - 7 (200 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 848641-69-0

  • Linear Formula C₁₀H₂₁N₂O₄P

  • Molecular Weight 264.26 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (4902940025)
density   1.15 g/cm3
pH   5 - 7 (200 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 98.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 0.1 %
  Water (K. F.): ≤ 1.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?