• USA Home
  • 4.90295 - 1-Ethyl-3-methylpyridinium perfluorobutanesulfonate

EMAIL THIS PAGE TO A FRIEND
4.90295 EMD Millipore

1-Ethyl-3-methylpyridinium perfluorobutanesulfonate

1-Ethyl-3-methylpyridinium perfluorobutanesulfonate for synthesis. CAS 1015420-87-7, molar mass 421.28 g/mol., for synthesis

  • CAS Number 1015420-87-7

  • Linear Formula C₁₂H₂₁F₉NO₃S

  • Molecular Weight 421.28 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (4902950025)
density   1.52 g/cm3 (20 °C)
mp   -6 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 98.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 0.1 %
  Water (K. F.): ≤ 1.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?