• USA Home
  • 4.91029 - 1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate

EMAIL THIS PAGE TO A FRIEND
4.91029 EMD Millipore

1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate

1-Butyl-1-methylpyrrolidinium trifluoromethanesulfonate high purity. CAS 367522-96-1, chemical formula C₉H₂₀N CF₃O₃S., high purity

  • CAS Number 367522-96-1

  • Linear Formula C₁₀H₂₀F₃NO₃S

  • Molecular Weight 291.34 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (4910290025)
density   1.25 g/cm3 (20 °C)
mp   3 °C
pH   6 (H₂O, 25 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (electrophoresis): ≥ 99.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 100 ppm
  Water (K. F.): ≤ 100 ppm

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?