• USA Home
  • 4.91061 - 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

EMAIL THIS PAGE TO A FRIEND
4.91061 EMD Millipore

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate high purity. CAS 145022-44-2, chemical formula C₆H₁₁N₂ CF₃O₃S. 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate MSDS (material safety data sheets) or SDS, storage and handling info here., high purity

  • CAS Number 145022-44-2

  • Linear Formula C₇H₁₁F₃N₂O₃S

  • Molecular Weight 260.24 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
density   1.386 g/cm3 (20 °C)
mp   -12 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 99.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 100 ppm
  Water (K. F.): ≤ 200 ppm

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?