• USA Home
  • 4.91078 - 1-Hexyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate

EMAIL THIS PAGE TO A FRIEND
4.91078 EMD Millipore

1-Hexyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate

1-Hexyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate high purity. CAS 713512-19-7, pH 6 (H₂O, 20 °C)., high purity

  • CAS Number 713512-19-7

  • Linear Formula C₁₆H₁₉F₁₈N₂P

  • Molecular Weight 612.29 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (4910780025)
density   1.56 g/cm3 (20 °C)
mp   -14 °C
pH   6 (H₂O, 20 °C)
viscosity kinematic   74.30 mm2/s (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 99.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 100 ppm
  Water (K. F.): ≤ 100 ppm

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?