• USA Home
  • 4.91232 - 1-Butyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate

EMAIL THIS PAGE TO A FRIEND
4.91232 EMD Millipore

1-Butyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate

1-Butyl-3-methylimidazolium tris(pentafluoroethyl)trifluorophosphate high purity. CAS 917762-91-5, pH 5 (H₂O, 20 °C) Aqueous solution., high purity

  • CAS Number 917762-91-5

  • Linear Formula C₁₄H₁₅F₁₈N₂P

  • Molecular Weight 584.23 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (4912320100)
density   1.63 g/cm3 (20 °C)
pH   5 (H₂O, 20 °C) Aqueous solution
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 99.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 100 ppm
  Water (K. F.): ≤ 100 ppm

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?