• USA Home
  • 4.92046 - 1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

EMAIL THIS PAGE TO A FRIEND
4.92046 EMD Millipore

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide ultra pure. CAS 223437-11-4, chemical formula C₉H₂₀N C₂F₆NO₄S₂., ultra pure

  • CAS Number 223437-11-4

  • Linear Formula C₁₁H₂₀F₆N₂O₄S₂

  • Molecular Weight 422.41 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (4920460025)
  500 g in Glass bottle (4920460500)
density   1.40 g/cm3
mp   -6 °C
pH   5 (H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (electrophoresis): ≥ 99.0 %
  Identity (NMR): passes test
  Halides (IC): ≤ 10 ppm
  Water (K. F.): ≤ 10 ppm

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2935 90 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?