• USA Home
  • 8.00390 - 2-Aminobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.00390 EMD Millipore

2-Aminobenzotrifluoride

2-Aminobenzotrifluoride for synthesis. CAS 88-17-5, chemical formula 2-(NH₂)C₆H₄CF₃., for synthesis

Synonym: α,α,α-Trifluoro-o-toluidine, 2-(Trifluoromethyl)aniline

  • CAS Number 88-17-5

  • Linear Formula C₇H₆F₃N

  • Molecular Weight 161.12 g/mol

  •  EC Number 201-806-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8003900100)
bp   170 - 172 °C (20 hPa)
vapor pressure   1.3 hPa
solubility   4 g/l
density   1.30 g/cm3 (20 °C)
flash point   56 °C
mp   -33 °C
LD 50 oral   LD50 Rat 480 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.294 - 1.296
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 43 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
H318: Causes serious eye damage.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat.
P273: Avoid release to the environment.
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2942 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2942 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2942 , 6.1, III
Categories of danger 
harmful, irritant, dangerous for the environment
R Phrase 
R 22-41-51/53
Harmful if swallowed.Risk of serious damage to eyes.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-39-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?