• USA Home
  • 8.00391 - 3-Aminobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
8.00391 EMD Millipore

3-Aminobenzotrifluoride

3-Aminobenzotrifluoride for synthesis. CAS No. 98-16-8, EC Number 202-643-4., for synthesis

Synonym: 3-(Trifluoromethyl)aniline, α,α,α-Trifluoro-m-toluidine

  • CAS Number 98-16-8

  • Linear Formula C₇H₆F₃N

  • Molecular Weight 161.13 g/mol

  •  EC Number 202-643-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   186 - 188 °C (1013 hPa)
vapor pressure   1.6 hPa (20 °C)
solubility   5 g/l
density   1.30 g/cm3 (20 °C)
flash point   91 °C
autoignition temp.   600 °C
mp   5 - 7 °C
LD 50 oral   LD50 Rat 480 mg/kg
LD 50 dermal   LD50 Rabbit 1330 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.294 - 1.297
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 43 00
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
H315: Causes skin irritation.
H318: Causes serious eye damage.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear eye protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
RTECS 
XU9180000
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2948 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2948 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2948 , 6.1, II
Categories of danger 
harmful, irritant, dangerous for the environment
R Phrase 
R 21/22-38-41-51/53
Harmful in contact with skin and if swallowed.Irritating to skin.Risk of serious damage to eyes.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-36/37/39-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.Avoid release to the environment. Refer to special instructions/ Safety data sheets.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?