• USA Home
  • 8.00479 - 4-Chloro-3-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.00479 EMD Millipore

4-Chloro-3-nitrobenzoic acid

4-Chloro-3-nitrobenzoic acid for synthesis. CAS 96-99-1, chemical formula 4-(Cl)-3-(NO₂)C₆H₃COOH., for synthesis

  • CAS Number 96-99-1

  • Linear Formula C₇H₄ClNO₄

  • Molecular Weight 201.56 g/mol

  •  EC Number 202-550-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8004790005)
density   1.645 g/cm3 (20 °C)
autoignition temp.   340 °C
mp   180 - 183 °C
bulk density   350 kg/m3
LD 50 oral   LD50 Rat 3150 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 180 °C
  Melting range (upper value): ≤ 183 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
DG5425050
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?