• USA Home
  • 8.00507 - 4-Bromobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.00507 EMD Millipore

4-Bromobenzoic acid

4-Bromobenzoic acid for synthesis. CAS 586-76-5, chemical formula 4-(Br)C₆H₄COOH., for synthesis

  • CAS Number 586-76-5

  • Linear Formula C₇H₅BrO₂

  • Molecular Weight 201.01 g/mol

  •  EC Number 209-581-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8005070050)
solubility   0.06 g/l
mp   252 - 255 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 252 °C
  Melting range (upper value): ≤ 257 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H302: Harmful if swallowed.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Hazard Symbols 
Harmful  Harmful
Categories of danger 
irritant
R Phrase 
R 36
Irritating to eyes.
S Phrase 
S 22-26
Do not breathe dust.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?