EMAIL THIS PAGE TO A FRIEND
8.00606 EMD Millipore

Ethyl propionate

Ethyl propionate for synthesis. CAS 105-37-3, EC Number 203-291-4, chemical formula CH₃CH₂COOC₂H₅., for synthesis

Synonym: Propionic acid ethyl ester

  • CAS Number 105-37-3

  • Linear Formula C₅H₁₀O₂

  • Molecular Weight 102.13 g/mol

  •  EC Number 203-291-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8006060100)
bp   99 °C (1013 hPa)
expl. lim.     1.8 - 11 %(V)
vapor pressure   26.7 hPa (20 °C)
solubility   17 g/l
density   0.89 g/cm3 (20 °C)
flash point   12 °C
autoignition temp.   475 °C
mp   -74 °C
pH   7 (H₂O, 20 °C)
LD 50 oral   LD50 Rat 8732 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.889 - 0.891
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 50 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P260: Do not breathe vapour.
P262: Do not get in eyes, on skin, or on clothing.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
RTECS 
UF3675000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1195 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1195 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1195 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 16-23-24-29-33
Keep away from sources of ignition - No smoking.Do not breathe vapour.Avoid contact with skin.Do not empty into drains.Take precautionary measures against static discharges.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?