EMAIL THIS PAGE TO A FRIEND
8.00607 EMD Millipore

Isophthalic acid

Isophthalic acid for synthesis. CAS 121-91-5, EC Number 204-506-4, chemical formula C₆H₄-1,3-(COOH)₂., for synthesis

Synonym: 1,3-Benzenedicarboxylic acid

  • CAS Number 121-91-5

  • Linear Formula C₈H₆O₄

  • Molecular Weight 166.13 g/mol

  •  EC Number 204-506-4

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   100 g in Plastic bottle (8006070100)
vapor pressure   6.1 hPa (250 °C)
solubility   0.12 g/l
density   1.54 g/cm3 (20 °C)
autoignition temp.   700 °C
mp   345 - 348 °C (sublimed)
LD 50 oral   LD50 Rat 10400 mg/kg
LD 50 dermal   LD50 Rabbit > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
RTECS 
NT2007000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?