• USA Home
  • 8.00615 - 2-Pyrazinecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.00615 EMD Millipore

2-Pyrazinecarboxylic acid

2-Pyrazinecarboxylic acid for synthesis. CAS 98-97-5, molar mass 124.09 g/mol., for synthesis

Synonym: Pyrazinoic acid

  • CAS Number 98-97-5

  • Linear Formula C₅H₄N₂O₂

  • Molecular Weight 124.09 g/mol

  •  EC Number 202-718-1

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   25 g in Glass bottle (8006150025)
mp   225 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 220 °C
  Melting range (upper value): ≤ 225 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?