• USA Home
  • 8.00616 - 2,3-Pyrazinedicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
8.00616 EMD Millipore

2,3-Pyrazinedicarboxylic acid

2,3-Pyrazinedicarboxylic acid for synthesis. CAS 89-01-0, molar mass 168.11 g/mol., for synthesis

  • CAS Number 89-01-0

  • Linear Formula C₆H₄N₂O₄

  • Molecular Weight 168.11 g/mol

  •  EC Number 201-875-3

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   25 g in Glass bottle (8006160025)
mp   188 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
RTECS 
UQ2500000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?