• USA Home
  • 8.00650 - 4-Nitrophthalic acid

EMAIL THIS PAGE TO A FRIEND
8.00650 EMD Millipore

4-Nitrophthalic acid

4-Nitrophthalic acid for synthesis. CAS 610-27-5, chemical formula 4-(NO₂)C₆H₃-1,2-(COOH)₂., for synthesis

Synonym: 4-Nitrobenzene-1,2-dicarboxylic acid

  • CAS Number 610-27-5

  • Linear Formula C₈H₅NO₆

  • Molecular Weight 211.13 g/mol

  •  EC Number 210-215-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8006500100)
solubility   880 g/l
mp   162 - 165 °C
pH   1 (10 g/l, H₂O, 20 °C)
bulk density   685 kg/m3
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 80.0 % (a/a)
  3-Nitrophthalic acid (GC, area%): ≤ 20.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?