EMAIL THIS PAGE TO A FRIEND
8.00660 EMD Millipore

Mucic acid

Mucic acid for synthesis. CAS 526-99-8, EC Number 208-404-0, chemical formula HOOCCH(OH)₄COOH., for synthesis

Synonym: Galactaric acid, Tetrahydroxyadipic acid

  • CAS Number 526-99-8

  • Linear Formula C₆H₁₀O₈

  • Molecular Weight 210.14 g/mol

  •  EC Number 208-404-0

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   100 g in Plastic bottle (8006600100)
solubility   3.3 g/l soluble
mp   220 - 225 °C (decomposition)
bulk density   400 kg/m3
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 19 98
RTECS 
LW5180000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?