EMAIL THIS PAGE TO A FRIEND
8.00663 EMD Millipore

Diethyl pimelate

Diethyl pimelate for synthesis. CAS 2050-20-6, chemical formula C₂H₅OOC(CH₂)₅COOC₂H₅., for synthesis

Synonym: Diethyl heptanedioate, Pimelic acid diethyl ester

  • CAS Number 2050-20-6

  • Linear Formula C₁₁H₂₀O₄

  • Molecular Weight 216.27 g/mol

  •  EC Number 218-083-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8006630025)
bp   252 - 253 °C
vapor pressure   0.004 hPa (25 °C)
solubility   1.97 g/l slightly soluble
density   0.99 g/cm3 (20 °C)
flash point   113 °C
mp   -24 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.993 - 0.995
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?