• USA Home
  • 8.00738 - 2,4-Dihydroxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.00738 EMD Millipore

2,4-Dihydroxybenzoic acid

2,4-Dihydroxybenzoic acid for synthesis. CAS 89-86-1, chemical formula 2,4-(OH)₂C₆H₃COOH., for synthesis

Synonym: ß-Resorcylic acid

  • CAS Number 89-86-1

  • Linear Formula C₇H₆O₄

  • Molecular Weight 154.12 g/mol

  •  EC Number 201-946-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8007380100)
solubility   8 g/l
bulk density   450 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?