EMAIL THIS PAGE TO A FRIEND
8.00884 EMD Millipore

Propyl formate

Propyl formate for synthesis. CAS No. 110-74-7, EC Number 203-798-0., for synthesis

Synonym: Formic acid propyl ester

  • CAS Number 110-74-7

  • Linear Formula C₄H₈O₂

  • Molecular Weight 88.1 g/mol

  •  EC Number 203-798-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8008840050)
bp   81 °C (1013 hPa)
expl. lim.     2.4 - 7.8 %(V)
vapor pressure   84 hPa (20 °C)
solubility   28 g/l (slow decomposition)
density   0.90 g/cm3 (20 °C)
flash point   -5 °C
autoignition temp.   450 °C
mp   -93 °C
LD 50 oral   LD50 Rat 3980 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.904 - 0.907
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 13 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H336: May cause drowsiness or dizziness.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P233: Keep container tightly closed.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1281 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1281 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1281 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-36/37-67
Highly flammable.Irritating to eyes and respiratory system.Vapours may cause drowsiness and dizziness.
S Phrase 
S 9-16-24-33
Keep container in a well-ventilated place.Keep away from sources of ignition - No smoking.Avoid contact with skin.Take precautionary measures against static discharges.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?