EMAIL THIS PAGE TO A FRIEND
8.00936 EMD Millipore

4-Ethylaniline

4-Ethylaniline for synthesis. CAS 589-16-2, chemical formula 4-(C₂H₅)C₆H₄NH₂., for synthesis

Synonym: 1-Amino-4-ethylbenzene

  • CAS Number 589-16-2

  • Linear Formula C₈H₁₁N

  • Molecular Weight 121.18 g/mol

  •  EC Number 209-637-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8009360100)
vapor pressure   1.1 hPa (50 °C)
density   0.97 g/cm3 (20 °C)
flash point   97 °C
mp   -5 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.967 - 0.969
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 1 slightly water endangering
Declaration (railroad and road) ADR, RID 
UN 1602 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 1602 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 1602 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?