EMAIL THIS PAGE TO A FRIEND
8.00937 EMD Millipore

2-Ethylaniline

2-Ethylaniline for synthesis. CAS 578-54-1, chemical formula 2-(C₂H₅)C₆H₄NH₂., for synthesis

Synonym: 1-Amino-2-ethylbenzene

  • CAS Number 578-54-1

  • Linear Formula C₈H₁₁N

  • Molecular Weight 121.18 g/mol

  •  EC Number 209-424-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8009370250)
bp   210 °C (1013 hPa)
vapor pressure   0.15 hPa (20 °C)
density   0.98 g/cm3 (20 °C)
flash point   92 °C
mp   -46 °C
LD 50 oral   LD50 Rat 1260 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.982 - 0.983
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H302: Harmful if swallowed.
RTECS 
BX9800000
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2273 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2273 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2273 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?