• USA Home
  • 8.00950 - Ethylene glycol diacetate

EMAIL THIS PAGE TO A FRIEND
8.00950 EMD Millipore

Ethylene glycol diacetate

Ethylene glycol diacetate for synthesis. CAS 111-55-7, EC Number 203-881-1, chemical formula CH₃COOCH₂CH₂OOCCH₃., for synthesis

Synonym: 1,2-Diacetoxyethane, Ethylene diacetate

  • CAS Number 111-55-7

  • Linear Formula C₆H₁₀O₄

  • Molecular Weight 146.14 g/mol

  •  EC Number 203-881-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8009500100)
bp   186 - 191 °C (1013 hPa)
vapor pressure   0.33 hPa (20 °C)
solubility   160 g/l
density   1.10 g/cm3 (20 °C)
flash point   88 °C
autoignition temp.   482 °C
mp   -31 °C
LD 50 oral   LD50 Rat 6850 mg/kg
LD 50 dermal   LD50 Rabbit 9328 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.105 - 1.108
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 39 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?