EMAIL THIS PAGE TO A FRIEND
8.00956 EMD Millipore

1-Ethylpiperidine

1-Ethylpiperidine for synthesis. CAS 766-09-6, pH 12.2 (50 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 766-09-6

  • Linear Formula C₇H₁₅N

  • Molecular Weight 113.2 g/mol

  •  EC Number 212-161-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8009560250)
expl. lim.     1.9 - 12.1 %(V)
vapor pressure   12.76 hPa (20 °C)
solubility   6.0 g/l
density   0.83 g/cm3 (20 °C)
flash point   17 °C
autoignition temp.   205 °C
mp   <-50 °C
pH   12.2 (50 g/l, H₂O, 20 °C)
viscosity kinematic   1.06 mm2/s (20 °C)
LD 50 oral   LD50 Rat 280 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.824 - 0.826
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H301 + H331: Toxic if swallowed or if inhaled
H312: Harmful in contact with skin.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
P240: Ground/bond container and receiving equipment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
P403 + P233: Store in a well-ventilated place. Keep container tightly closed.
RTECS 
TN0250000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 2386 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 2386 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2386 , 3 (8), II, Segregation Group: 18 (Alkalis)
Categories of danger 
highly flammable, harmful, corrosive
R Phrase 
R 11-20/21/22-34
Highly flammable.Harmful by inhalation, in contact with skin and if swallowed.Causes burns.
S Phrase 
S 16-26-36/37/39-45
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?