EMAIL THIS PAGE TO A FRIEND
8.01110 EMD Millipore

3-Aminopyridine

3-Aminopyridine for synthesis. CAS No. 462-08-8, EC Number 207-322-2., for synthesis

Synonym: 3-Pyridylamine

  • CAS Number 462-08-8

  • Linear Formula C₅H₆N₂

  • Molecular Weight 94.11 g/mol

  •  EC Number 207-322-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8011100025)
vapor pressure   1 hPa (25 °C)
solubility   >1000 g/l
flash point   88 °C
autoignition temp.   628 °C
mp   60 - 63 °C
pH   10 (100 g/l, H₂O, 20 °C)
bulk density   500 kg/m3
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 60 °C
  Melting range (upper value): ≤ 63 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H301: Toxic if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2671 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2671 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2671 , 6.1, II, Segregation Group: 18 (Alkalis)
Categories of danger 
toxic, irritant
R Phrase 
R 25-36/38
Toxic if swallowed.Irritating to eyes and skin.
S Phrase 
S 37-45
Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?