• USA Home
  • 8.01241 - Ethyl 2-ethoxy-1,2-dihydro-1-quinolinecarboxylate

EMAIL THIS PAGE TO A FRIEND
8.01241 EMD Millipore

Ethyl 2-ethoxy-1,2-dihydro-1-quinolinecarboxylate

Ethyl 2-ethoxy-1,2-dihydro-1-quinolinecarboxylate for synthesis. CAS 16357-59-8, molar mass 247.29 g/mol., for synthesis

  • CAS Number 16357-59-8

  • Linear Formula C₁₄H₁₇NO₃

  • Molecular Weight 247.29 g/mol

  •  EC Number 240-418-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8012410010)
bp   125 - 128 °C (0.13 hPa)
mp   63 - 66 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 99.0 %
  Melting range (lower value): ≥ 63 °C
  Melting range (upper value): ≤ 66 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 49 90
RTECS 
VB2010000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?