• USA Home
  • 8.01265 - Diethyl benzylmalonate

EMAIL THIS PAGE TO A FRIEND
8.01265 EMD Millipore

Diethyl benzylmalonate

Diethyl benzylmalonate for synthesis. CAS 607-81-8, chemical formula C₆H₅CH₂CH(COOC₂H₅)₂., for synthesis

Synonym: Benzylmalonic acid diethyl ester

  • CAS Number 607-81-8

  • Linear Formula C₁₄H₁₈O₄

  • Molecular Weight 250.29 g/mol

  •  EC Number 210-142-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8012650025)
bp   162 °C (13 hPa)
density   1.07 g/cm3 (20 °C)
flash point   >110 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.072 - 1.074
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?