EMAIL THIS PAGE TO A FRIEND
8.01270 EMD Millipore

2-Aminopyrimidine

2-Aminopyrimidine for synthesis. CAS 109-12-6, molar mass 95.1 g/mol., for synthesis

Synonym: 2-Pyrimidinylamine

  • CAS Number 109-12-6

  • Linear Formula C₄H₅N₃

  • Molecular Weight 95.1 g/mol

  •  EC Number 203-648-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8012700100)
bp   158 - 160 °C (186 hPa)
flash point   103 °C
mp   122 - 126 °C
bulk density   650 kg/m3
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 122 °C
  Melting range (upper value): ≤ 126 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
RTECS 
UV6326000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?