• USA Home
  • 8.01274 - Diethyl butylmalonate

EMAIL THIS PAGE TO A FRIEND
8.01274 EMD Millipore

Diethyl butylmalonate

Diethyl butylmalonate for synthesis. CAS 133-08-4, chemical formula CH₃(CH₂)₃CH(COOC₂H₅)₂., for synthesis

Synonym: Butylmalonic acid diethyl ester

  • CAS Number 133-08-4

  • Linear Formula C₁₁H₂₀O₄

  • Molecular Weight 216.27 g/mol

  •  EC Number 205-089-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8012740250)
bp   235 - 240 °C (1013 hPa)
density   0.975 g/cm3 (20 °C)
flash point   113 °C DIN 51758
LD 50 oral   LD50 Rat > 10000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.974 - 0.976
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?