EMAIL THIS PAGE TO A FRIEND
8.01300 EMD Millipore

Ethyl pivalate

Ethyl pivalate for synthesis. CAS 3938-95-2, EC Number 223-520-1, chemical formula (CH₃)₃CCOOC₂H₅., for synthesis

Synonym: Pivalic acid ethyl ester, Ethyl trimethylacetate, Trimethylacetic acid ethyl ester, 2,2-Dimethylpropionic acid ethyl ester

  • CAS Number 3938-95-2

  • Linear Formula C₇H₁₄O₂

  • Molecular Weight 130.18 g/mol

  •  EC Number 223-520-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8013000100)
bp   118 °C (1013 hPa)
density   0.86 g/cm3 (20 °C)
flash point   12 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.854 - 0.856
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 60 90
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 3272 , 3, II
Declaration (transport by air) IATA-DGR 
UN 3272 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 3272 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 9-16
Keep container in a well-ventilated place.Keep away from sources of ignition - No smoking.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?