• USA Home
  • 8.01303 - 3-Morpholino propylamine

EMAIL THIS PAGE TO A FRIEND
8.01303 EMD Millipore

3-Morpholino propylamine

3-Morpholino propylamine for synthesis. CAS 123-00-2, pH 12 (H₂O, 20 °C)., for synthesis

Synonym: N-(3-Aminopropyl)morpholine, 4-(3-Aminopropyl)morpholine

  • CAS Number 123-00-2

  • Linear Formula C₇H₁₆N₂O

  • Molecular Weight 144.22 g/mol

  •  EC Number 204-590-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   224 °C (1013 hPa)
vapor pressure   <1 hPa (20 °C)
solubility   1000 g/l
density   0.99 g/cm3 (20 °C)
flash point   100 °C
autoignition temp.   250 °C
mp   -15 °C
pH   12 (H₂O, 20 °C)
LD 50 oral   LD50 Rat 1790 mg/kg
LD 50 dermal   LD50 Rabbit 2300 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97 %
  Density (d 20 °C/ 4 °C): 0.984 - 0.986
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Hazard Statement(s) 
H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2735 , 8, II
Declaration (transport by air) IATA-DGR 
UN 2735 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 2735 , 8, II, Segregation Group: 18 (Alkalis)
Categories of danger 
harmful, corrosive
R Phrase 
R 22-34
Harmful if swallowed.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?