EMAIL THIS PAGE TO A FRIEND
8.01305 EMD Millipore

Dodecanedioic acid

Dodecanedioic acid for synthesis. CAS 693-23-2, chemical formula HOOC(CH₂)₁₀COOH., for synthesis

Synonym: 1,12-Decanedicarboxylic acid

  • CAS Number 693-23-2

  • Linear Formula C₁₂H₂₂O₄

  • Molecular Weight 230.3 g/mol

  •  EC Number 211-746-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8013050100)
bp   205 °C (1 hPa)
vapor pressure   <0.01 hPa (20 °C)
solubility   0.04 g/l
flash point   220 °C
autoignition temp.   390 °C
mp   126 - 129 °C
pH   5 (H₂O, 20 °C) (aqueous suspension)
bulk density   600 kg/m3
LD 50 oral   LD50 Rat > 3000 mg/kg
LD 50 dermal   LD50 Rabbit > 6000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 126 °C
  Melting range (upper value): ≤ 129 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Hazard Statement(s) 
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
NWG not water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36
Irritating to eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?