• USA Home
  • 8.01312 - 2-Amino-5-chlorobenzophenone

EMAIL THIS PAGE TO A FRIEND
8.01312 EMD Millipore

2-Amino-5-chlorobenzophenone

2-Amino-5-chlorobenzophenone for synthesis. CAS 719-59-5, chemical formula 2-(NH₂)-5-(Cl)C₆H₃COC₆H₅., for synthesis

Synonym: 5-Chloro-2-aminobenzophenone

  • CAS Number 719-59-5

  • Linear Formula C₁₃H₁₀ClNO

  • Molecular Weight 231.68 g/mol

  •  EC Number 211-949-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Plastic bottle (8013120250)
bp   207 °C (1013 hPa)
flash point   211 °C
mp   97 - 99 °C
LD 50 oral   LD50 Rat 10000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 95 °C
  Melting range (upper value): ≤ 99 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 39 00
RTECS 
PC4933500
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?