EMAIL THIS PAGE TO A FRIEND
8.01338 EMD Millipore

2-Aminopyrazine

2-Aminopyrazine for synthesis. CAS 5049-61-6, molar mass 95.1 g/mol., for synthesis

  • CAS Number 5049-61-6

  • Linear Formula C₄H₅N₃

  • Molecular Weight 95.1 g/mol

  •  EC Number 225-748-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8013380010)
mp   117 - 120 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 117 °C
  Melting range (upper value): ≤ 120 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?