EMAIL THIS PAGE TO A FRIEND
8.01339 EMD Millipore

1-Adamantanol

1-Adamantanol for synthesis. CAS 768-95-6, molar mass 152.23 g/mol., for synthesis

Synonym: 1-Hydroxyadamantane

  • CAS Number 768-95-6

  • Linear Formula C₁₀H₁₆O

  • Molecular Weight 152.23 g/mol

  •  EC Number 212-202-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8013390010)
mp   246 - 248 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 19 00
RTECS 
AU4980000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?