EMAIL THIS PAGE TO A FRIEND
8.01418 EMD Millipore

4-Aminobenzamide

4-Aminobenzamide for synthesis. CAS 2835-68-9, chemical formula 4-(NH₂)C₆H₄CONH₂., for synthesis

Synonym: 4-Aminobenzoic acid amide

  • CAS Number 2835-68-9

  • Linear Formula C₇H₈N₂O

  • Molecular Weight 136.15 g/mol

  •  EC Number 220-612-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8014180100)
mp   180 - 183 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 180 °C
  Melting range (upper value): ≤ 183 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Hazard Statement(s) 
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37
Irritating to eyes and respiratory system.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?