• USA Home
  • 8.01419 - 2-Aminobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.01419 EMD Millipore

2-Aminobenzonitrile

2-Aminobenzonitrile for synthesis. CAS 1885-29-6, chemical formula 2-(NH₂)C₆H₄CN., for synthesis

Synonym: Anthranilic acid nitrile, Anthranilonitrile, 2-Cyananiline

  • CAS Number 1885-29-6

  • Linear Formula C₇H₆N₂

  • Molecular Weight 118.13 g/mol

  •  EC Number 217-549-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8014190100)
  25 g in Glass bottle (8014190025)
density   1.11 g/cm3 (50 °C)
flash point   145 °C
autoignition temp.   >200 °C DIN 51794
mp   46 - 49 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 47 °C
  Melting range (upper value): ≤ 50 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?