EMAIL THIS PAGE TO A FRIEND
8.01518 EMD Millipore

2-BENZOYLPYRIDINE

2-BENZOYLPYRIDINE FOR SYNTHESIS. CAS 91-02-1, molar mass 183.2 g/mol., FOR SYNTHESIS

Synonym: 2-Benzoylpyridine

  • CAS Number 91-02-1

  • Linear Formula C₁₂H₉NO

  • Molecular Weight 183.2 g/mol

  •  EC Number 202-034-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Glass bottle (8015180250)
bp   170 - 172 °C (20 hPa)
vapor pressure   10 hPa (170 °C)
flash point   150 °C
mp   42 - 45 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 42 °C
  Melting range (upper value): ≤ 45 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
RTECS 
OB6400000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?