EMAIL THIS PAGE TO A FRIEND
8.01521 EMD Millipore

Benzylacetone

Benzylacetone for synthesis. CAS No. 2550-26-7, EC Number 219-847-4., for synthesis

Synonym: 4-Phenyl-2-butanone

  • CAS Number 2550-26-7

  • Linear Formula C₁₀H₁₂O

  • Molecular Weight 148.2 g/mol

  •  EC Number 219-847-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8015210250)
vapor pressure   0.0386 hPa (20 °C)
density   0.99 g/cm3 (20 °C)
flash point   101 °C
LD 50 oral   LD50 Rat 3200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.986 - 0.988
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 39 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?