EMAIL THIS PAGE TO A FRIEND
8.01524 EMD Millipore

2-Benzylpyridine

2-Benzylpyridine for synthesis. CAS 101-82-6, chemical formula 2-(C₆H₅CH₂)C₅H₄N., for synthesis

Synonym: 2-(Phenylmethyl)-pyridine

  • CAS Number 101-82-6

  • Linear Formula C₁₂H₁₁N

  • Molecular Weight 169.22 g/mol

  •  EC Number 202-979-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8015240100)
density   1.05 g/cm3 (20 °C)
mp   8 - 10 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.053 - 1.057
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
RTECS 
US2620000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?