EMAIL THIS PAGE TO A FRIEND
8.01525 EMD Millipore

4-Benzylpyridine

4-Benzylpyridine for synthesis. CAS 2116-65-6, chemical formula 4-(C₆H₅CH₂)C₅H₄N., for synthesis

Synonym: 4-(Phenylmethyl)-pyridine

  • CAS Number 2116-65-6

  • Linear Formula C₁₂H₁₁N

  • Molecular Weight 169.22 g/mol

  •  EC Number 218-319-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8015250100)
bp   287 °C (1013 hPa)
vapor pressure   10 hPa (135 °C)
density   1.06 g/cm3 (20 °C)
flash point   115 °C
mp   9 - 11 °C
pH   8 (H₂O, 20 °C)
LD 50 oral   LD50 Rat 593 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.061 - 1.063
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-38
Harmful if swallowed.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?