• USA Home
  • 8.01535 - Bromodiphenylmethane

EMAIL THIS PAGE TO A FRIEND
8.01535 EMD Millipore

Bromodiphenylmethane

Bromodiphenylmethane for synthesis. CAS No. 776-74-9, EC Number 212-279-8., for synthesis

Synonym: Benzhydryl bromide, Diphenylbromomethane, Diphenylmethyl bromide

  • CAS Number 776-74-9

  • Linear Formula C₁₃H₁₁Br

  • Molecular Weight 247.13 g/mol

  •  EC Number 212-279-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8015350100)
bp   184 °C (25 hPa)
solubility   0.45 g/l
flash point   112 °C
mp   35 - 40 °C
pH   2 (H₂O, 20 °C) (saturated aqueous solution)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Melting range (lower value): ≥ 36 °C
  Melting range (upper value): ≤ 39 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H290: May be corrosive to metals.
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 1770 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1770 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1770 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
corrosive
R Phrase 
R 34-37
Causes burns.Irritating to respiratory system.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?