EMAIL THIS PAGE TO A FRIEND
8.01540 EMD Millipore

sec-Butylamine

sec-Butylamine for synthesis. CAS 13952-84-6, EC Number 237-732-7, chemical formula CH₃CH₂CH(NH₂)CH₃., for synthesis

Synonym: 2-Aminobutane

  • CAS Number 13952-84-6

  • Linear Formula C₄H₁₁N

  • Molecular Weight 73.14 g/mol

  •  EC Number 237-732-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8015400100)
bp   63 - 64 °C (1013 hPa)
expl. lim.     1.8 - 9.0 %(V)
vapor pressure   182 hPa (20 °C)
solubility   112 g/l
density   0.72 g/cm3 (20 °C)
flash point   -20 °C
autoignition temp.   290 °C
mp   -104 °C
LD 50 oral   LD50 Rat 152 mg/kg
LD 50 dermal   LD50 Rabbit 2500 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.722 - 0.723
  Water (K. F.): ≤ 1.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 19 99
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H332: Harmful if inhaled.
H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
EO3325000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 2733 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 2733 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2733 , 3 (8), II, Segregation Group: 18 (Alkalis)
Categories of danger 
highly flammable, harmful, corrosive, dangerous for the environment
R Phrase 
R 11-20/22-35-50
Highly flammable.Harmful by inhalation and if swallowed.Causes severe burns.Very toxic to aquatic organisms.
S Phrase 
S 9-16-26-28-36/37/39-45-61
Keep container in a well-ventilated place.Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.After contact with skin, wash immediately with plenty of soap and water.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?