EMAIL THIS PAGE TO A FRIEND
8.01675 EMD Millipore

2-Bromomesitylene

2-Bromomesitylene for synthesis. CAS 576-83-0, chemical formula 2-(Br)C₆H₂-1,3,5-(CH₃)₃., for synthesis

Synonym: 2-Bromo-1,3,5-trimethylbenzene

  • CAS Number 576-83-0

  • Linear Formula C₉H₁₁Br

  • Molecular Weight 199.08 g/mol

  •  EC Number 209-405-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8016750025)
bp   225 °C (1033 hPa)
density   1.32 g/cm3 (20 °C)
flash point   96 °C
mp   2 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.322 - 1.325
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 37
Irritating to respiratory system.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?