• USA Home
  • 8.01681 - N-Methylbenzylamine

EMAIL THIS PAGE TO A FRIEND
8.01681 EMD Millipore

N-Methylbenzylamine

N-Methylbenzylamine for synthesis. CAS 103-67-3, EC Number 203-133-4, chemical formula C₆H₅CH₂NHCH₃., for synthesis

Synonym: N-Benzylmethylamine

  • CAS Number 103-67-3

  • Linear Formula C₈H₁₁N

  • Molecular Weight 121.18 g/mol

  •  EC Number 203-133-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8016810250)
bp   184 - 190 °C (1013 hPa)
expl. lim.     1.4 - 7.7 %(V)
vapor pressure   2.1 hPa (20 °C)
solubility   65 g/l
density   0.94 g/cm3 (20 °C)
flash point   61 °C
autoignition temp.   380 °C
mp   -55 °C
pH   10 (10 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.941 - 0.943
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H290: May be corrosive to metals.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2735 , 8, II
Declaration (transport by air) IATA-DGR 
UN 2735 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 2735 , 8, II, Segregation Group: 18 (Alkalis)
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?