EMAIL THIS PAGE TO A FRIEND
8.01755 EMD Millipore

Butyl benzoate

Butyl benzoate for synthesis. CAS No. 136-60-7, EC Number 205-252-7., for synthesis

Synonym: Benzoic acid butyl ester

  • CAS Number 136-60-7

  • Linear Formula C₁₁H₁₄O₂

  • Molecular Weight 178.23 g/mol

  •  EC Number 205-252-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8017550100)
bp   250 °C (1013 hPa)
vapor pressure   0.2 hPa (20 °C)
solubility   0.06 g/l
density   1.01 g/cm3 (20 °C)
flash point   106 °C
autoignition temp.   440 °C
mp   -22 °C
LD 50 oral   LD50 Rat 735 mg/kg
LD 50 dermal   LD50 Rabbit 4000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.005 - 1.006
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 31 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
DG4925000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-36/38
Harmful if swallowed.Irritating to eyes and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?