EMAIL THIS PAGE TO A FRIEND
8.01813 EMD Millipore

Ethyl benzoate

Ethyl benzoate for synthesis. CAS 93-89-0, EC Number 202-284-3, chemical formula C₆H₅COOC₂H₅., for synthesis

Synonym: Benzoic acid ethyl ester

  • CAS Number 93-89-0

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.17 g/mol

  •  EC Number 202-284-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8018130250)
bp   213 - 215 °C (1013 hPa)
expl. lim.     1 %(V)
vapor pressure   0.24 hPa (20 °C)
solubility   0.5 g/l
density   1.05 g/cm3 (20 °C)
flash point   88 °C
autoignition temp.   490 °C
mp   -35 °C
LD 50 oral   LD50 Rat 2100 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.046 - 1.047
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 31 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Categories of danger 
irritant
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?