• USA Home
  • 8.01822 - 1-Benzyl-4-piperidinone

EMAIL THIS PAGE TO A FRIEND
8.01822 EMD Millipore

1-Benzyl-4-piperidinone

1-Benzyl-4-piperidinone for synthesis. CAS 3612-20-2, molar mass 189.25 g/mol., for synthesis

Synonym: N-Benzyl-4-piperidone

  • CAS Number 3612-20-2

  • Linear Formula C₁₂H₁₅NO

  • Molecular Weight 189.25 g/mol

  •  EC Number 222-782-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8018220050)
bp   248 °C (1013 hPa)
solubility   12 g/l
density   1.06 g/cm3 (20 °C)
flash point   71 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.058 - 1.060
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-36/38
Harmful if swallowed.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?