• USA Home
  • 8.01935 - 4-Bromopyridinium chloride

EMAIL THIS PAGE TO A FRIEND
8.01935 EMD Millipore

4-Bromopyridinium chloride

4-Bromopyridinium chloride for synthesis. CAS 19524-06-2, chemical formula C₅H₄BrN*HCl., for synthesis

Synonym: 4-Bromopyridine hydrochloride

  • CAS Number 19524-06-2

  • Linear Formula C₅H₅BrClN

  • Molecular Weight 194.46 g/mol

  •  EC Number 243-128-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8019350005)
mp   235 - 238 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, III
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?