EMAIL THIS PAGE TO A FRIEND
8.01959 EMD Millipore

Biphenyl-4-ol

Biphenyl-4-ol for synthesis. CAS No. 92-69-3, EC Number 202-179-2., for synthesis

  • CAS Number 92-69-3

  • Linear Formula C₁₂H₁₀O

  • Molecular Weight 170.21 g/mol

  •  EC Number 202-179-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8019590100)
bp   305 - 308 °C (1013 hPa)
solubility   0.7 g/l
flash point   160 °C
autoignition temp.   >250 °C
mp   162 - 165 °C
pH   7 (0.7 g/l, H₂O, 20 °C)
bulk density   550 kg/m3
LD 50 oral   LD50 Rat > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 162 °C
  Melting range (upper value): ≤ 165 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 19 90
Hazard Statement(s) 
H315: Causes skin irritation.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3077 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3077 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3077 , 9, III, Marine Pollutant: P
Categories of danger 
irritant, dangerous for the environment
R Phrase 
R 38-51/53
Irritating to skin.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?